N-Methyl-2-pyrrolidon (NMP) ≥99%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
A12260.0C
A12260.0E
A12260.36
A12260.0CEA
276
EUR
A12260.0C
A12260.0E
A12260.36
N-Methyl-2-pyrrolidon (NMP) ≥99%
N-Methyl-2-pyrrolidon (NMP)
Purity: 99+%
CAS No.: 872-50-4
Molecular Formula: C5H9NO
Formula Weight: 99.13
Flash Point: 86°(187°F)
Refractive Index: 1.4705
Odour: Amine-like
Melting Point: -24°
Boiling Point: 204°
Beilstein Reference: 106420
Merck Reference: 14,6117
MDL No.: MFCD00003193
Formule:
C₅H₉NO Kookpunt: 204 °C Smeltpunt: –24 °C Dichtheid: 1,033 Vlampunt: 86 °C (187 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00003193 CAS nummer: 872-50-4 EINECS: 212-828-1 Merck Index: 14,6117 |