1-Ethylpiperidine 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

118885000 118880025 118880100
ACRO118885000EA 75 EUR
ACRO118885000 ACRO118880025 ACRO118880100
1-Ethylpiperidine 99%
1-Ethylpiperidine
Formule: C₇H₁₅N
Kookpunt: 129…131 °C (760 mmHg)
Smeltpunt: –20 °C
Dichtheid: 0,824 g/cm³
Vlampunt: 17 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006507
CAS nummer: 766-09-6
EINECS: 212-161-6
UN: 2386
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4430 to 1.4450 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR