1-Methylpiperidine

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

KM07798.1 KM07798.10
MAYBKM07798.1EA 0 EUR
MAYBKM07798.1 MAYBKM07798.10
1-Methylpiperidine
1-Methylpiperidine
Formule: C₆H₁₃N
Kookpunt: 106,5 °C (1013 hPa)
Smeltpunt: –69 °C
Dichtheid: 0,82 g/cm³ (20 °C)
Vlampunt: 3 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00006491
CAS nummer: 626-67-5
EINECS: 210-959-9
UN: 2399
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR