Azulene ≥99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Bicyclo[5.3.0]decapentaene
L08271.03 L08271.MD L08271.06
L08271.03EA 171 EUR
L08271.03 L08271.MD L08271.06
Azulene ≥99%
Azulene
Purity: 99% CAS No.: 275-51-4 Molecular Formula: C10H8 Formula Weight: 128.17 Melting Point: 98-101° Boiling Point: 241-243° Merck Reference: 14,926 MDL No.: MFCD00003810
Formule: C₁₀H₈
Kookpunt: 241…243 °C
Smeltpunt: 98…101 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003810
CAS nummer: 275-51-4
EINECS: 205-993-6
UN: 3077
ADR: 9,III
Merck Index: 14,926