5-Methyl-1,3-benzodioxole

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 5-Methylbenzo[d][1,3]dioxole, 3,4-(Methylenedioxy)toluene

SB00943.1 SB00943EA SB00943.50
MAYBSB00943.1EA 0 EUR
MAYBSB00943.1 MAYBSB00943EA MAYBSB00943.50
5-Methyl-1,3-benzodioxole
5-Methyl-1,3-benzodioxole
Formule: C₈H₈O₂
MW: 136.15 g/mol
MDL Nummer: MFCD00005833
CAS nummer: 7145-99-5
EINECS: 230-453-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR