3-Cyanophenylboronic acid
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
AC35903.250 AC35903.1
MAYBAC35903.250EA 0 EUR
MAYBAC35903.250 MAYBAC35903.1
3-Cyanophenylboronic acid
3-Cyanophenylboronic acid
Formule: C₇H₆BNO₂
MDL Nummer: MFCD01318967
CAS nummer: 150255-96-2