2,6-Dibromo-4-chloroaniline
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
4-Chloro-2,6-dibromoaniline, 1-Amino-4-chloro-2,6-dibromobenzene
RDP00220.1
RDP00220.10
RDP00220.50
MAYBRDP00220.1EA
0
EUR
MAYBRDP00220.1
MAYBRDP00220.10
MAYBRDP00220.50
2,6-Dibromo-4-chloroaniline
2,6-Dibromo-4-chloroaniline
Formule:
C₆H₄Br₂ClN MW: 285.37 g/mol Kookpunt: 301 Smeltpunt: 96 |
MDL Nummer:
MFCD00051750 CAS nummer: 874-17-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...