2-Bromobenzoyl chloride

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 2-Bromobenzene-1-carbonyl chloride, o-Bromobenzoyl chloride

RDP00157.1 RDP00157.10 RDP00157.50
MAYBRDP00157.1EA 0 EUR
MAYBRDP00157.1 MAYBRDP00157.10 MAYBRDP00157.50
2-Bromobenzoyl chloride
2-Bromobenzoyl chloride
Formule: C₇H₄BrClO
Kookpunt: 245 °C (1013 hPa)
Smeltpunt: 11 °C
Dichtheid: 1,67 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000655
CAS nummer: 7154-66-7
EINECS: 230-507-4
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR