2-Bromobenzamide

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

RDP00161.1 RDP00161.50
MAYBRDP00161.1EA 0 EUR
MAYBRDP00161.1 MAYBRDP00161.50
2-Bromobenzamide
2-Bromobenzamide
Formule: C₇H₆BrNO
MW: 200.03 g/mol
Kookpunt: 198
Smeltpunt: 160…162 °C
Dichtheid: 1.609 g/cm³ (20 °C)
MDL Nummer: MFCD00007969
CAS nummer: 4001-73-4
EINECS: 223-650-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR