2-Bromobenzhydrazide ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

L00937.06 L00937.14
L00937.06EA 38.7 EUR
L00937.06 L00937.14
2-Bromobenzhydrazide ≥98%
2-Bromobenzhydrazide

Purity: 98+% CAS No.: 29418-67-5 Molecular Formula: C7H7BrN2O Formula Weight: 215.05 Melting Point: 152-154° Beilstein Reference: 1941021

Formule: C₇H₇BrN₂O
Smeltpunt: 152…154 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00051577
CAS nummer: 29418-67-5
EINECS: 249-614-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR