Volumetric flasks, class A, trapezoidal shape, BLAUBRAND®
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2.36408 2.36403 2.36404 2.36406 2.36407 2.36402 2.36401
BRND2.36408EA 52.4 EUR
BRND2.36408 BRND2.36403 BRND2.36404 BRND2.36406 BRND2.36407 BRND2.36402 BRND2.36401
Volumetric flasks, class A, trapezoidal shape, BLAUBRAND®
Kolven Maatkolven
Borosilicate glass 3.3, with PP stopper.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast blue enamel
  • DE-M marking
  • Total order from one batch, batch certificate included
*Wide neck.
Certificaten: DIN EN ISO 1042