2-Bromo-4-fluoroaniline
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
DP01928.1
DP01928.10
DP01928.50
MAYBDP01928.1EA
0
EUR
MAYBDP01928.1
MAYBDP01928.10
MAYBDP01928.50
2-Bromo-4-fluoroaniline
2-Bromo-4-fluoroaniline
Formule:
C₆H₅BrFN Kookpunt: 46…47 °C (0,05 mmHg) Smeltpunt: 41 °C Dichtheid: 1,67 g/cm³ (20 °C) |
MDL Nummer:
MFCD00042462 CAS nummer: 1003-98-1 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...