Veratraldehyde

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

9651.1 9651.2
ROTH9651.1EA 0 EUR
ROTH9651.1 ROTH9651.2
Veratraldehyde
Veratraldehyde
Formule: C₉H₁₀O₃
Kookpunt: 281 °C (1013 hPa)
Smeltpunt: 40…43 °C
Vlampunt: >100 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003363
CAS nummer: 120-14-9
EINECS: 204-373-2
Merck Index: 12,10084

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR