4-(Methylthio)benzaldehyde ≥97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B25427.14 B25427.22
B25427.14EA 51.9 EUR
B25427.14 B25427.22
4-(Methylthio)benzaldehyde ≥97%
4-(Methylthio)benzaldehyde
Purity: 97% CAS No.: 3446-89-7 Molecular Formula: C8H8OS Formula Weight: 152.22 Flash Point: 110°(230°F) Refractive Index: 1.6490 Melting Point: 6° Boiling Point: 118-121°/5mm Beilstein Reference: 636158 MDL No.: MFCD00006948
Formule: C₈H₈OS
Kookpunt: 118…121 °C (5 mmHg)
Smeltpunt: 6 °C
Dichtheid: 1,144
Vlampunt: >110 °C (230 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006948
CAS nummer: 3446-89-7
EINECS: 222-365-7
UN: 3334
ADR: 9,