Terephthalaldehyde 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
137891000 137895000
ACRO137891000EA 54.7 EUR
ACRO137891000 ACRO137895000
Terephthalaldehyde 98%
Terephthalaldehyde
Formule: C₈H₆O₂
Kookpunt: 245 °C (1013 hPa)
Smeltpunt: 114…116 °C
Dichtheid: 1,381 g/cm³ (20 °C)
Vlampunt: 76 °C
MDL Nummer: MFCD00006949
CAS nummer: 623-27-8
EINECS: 210-784-8
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 114 to 116 °C
GC ≥97.5 %
Specific optical rotation +35° to +39° (20 °C, 589 nm) (C=5, 1N HCl)