2,6-Dichlorobenzoyl chloride 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 2,6-Dichlorobenzoyl Chloride
182060100 182060500
ACRO182060100EA 48.1 EUR
ACRO182060100 ACRO182060500
2,6-Dichlorobenzoyl chloride 99%
2,6-Dichlorobenzoyl chloride
Formule: C₇H₃Cl₃O
Kookpunt: 142…143 °C (21 mmHg)
Smeltpunt: 18 °C
Dichtheid: 1,45 g/cm³
Vlampunt: >110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000662
CAS nummer: 4659-45-4
EINECS: 225-102-4
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow to light pink Liquid
Infrared spectrum Conforms
Refractive index 1.5590 to 1.5610 (20°C, 589 nm)
GC ≥98.5 %
Titration RCOCl ≥98.5 %