Chemical resistant overalls, Body Tech T
Leverancier: BEST FRANCE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
01074T030XL 01074T030XXL 01074T030XXXL
BESF01074T030XLEA 0 EUR
BESF01074T030XL BESF01074T030XXL BESF01074T030XXXL
Chemical resistant overalls, Body Tech T
Wegwerpkleding
These hooded overalls made from laminated PE provide protection against light splashes and sprays of hazardous liquids as well as against hazardous dusts.
  • Elastic cuffs, ankles and back
  • Good moisture wicking properties
  • Zip closure (double slider) with adhesive flap
  • Waterproof and impervious to certain solvents or alcohols
  • Highly resistant seams with thermally bonded tape
  • Anti-static according to EN 1149-5
Weight: 58 g/m²
Certificaten: Type 5/6, EN 1149-5; Cat. III