3,5-Dichlorobenzoyl chloride
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SB01888.1
SB01888.10
SB01888.50
MAYBSB01888.1EA
0
EUR
MAYBSB01888.1
MAYBSB01888.10
MAYBSB01888.50
3,5-Dichlorobenzoyl chloride
3,5-Dichlorobenzoyl chloride
Formule:
C₇H₃Cl₃O Kookpunt: 249 °C (1013 hPa) Smeltpunt: 26…28 °C Dichtheid: 1,478 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00009817 CAS nummer: 2905-62-6 EINECS: 220-813-6 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...