3,4-Dichlorobenzamide
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TL00384.1
TL00384.10
MAYBTL00384.1EA
0
EUR
MAYBTL00384.1
MAYBTL00384.10
3,4-Dichlorobenzamide
3,4-Dichlorobenzamide
Formule:
C₇H₅Cl₂NO |
MDL Nummer:
MFCD00053009 CAS nummer: 2670-38-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...