3,5-Dichlorobenzoic acid ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D0341-25G D0341-500G
TCIAD0341-25GEA 30.8 EUR
TCIAD0341-25G TCIAD0341-500G
3,5-Dichlorobenzoic acid ≥98.0% (by titrimetric analysis)
3,5-Dichlorobenzoic acid
Formule: C₇H₄Cl₂O₂
Kookpunt: 309 °C (1013 hPa)
Smeltpunt: 183…185 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002494
CAS nummer: 51-36-5
EINECS: 200-092-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=999fc8dc871c56e851bcc913c269fc0458fa0d53&vid=ac3605f0381b7b1803aec39c6e662aaa9a76fa78&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 185.0 to 189.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR