Autosampler syringes for CTC PAL3 systems
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8010-0164 8010-0130 8010-0138 8010-0163 8010-0168 8010-0169 8010-0171 8010-0173 8010-0174
HEWL8010-0164EA 133 EUR
HEWL8010-0164 HEWL8010-0130 HEWL8010-0138 HEWL8010-0163 HEWL8010-0168 HEWL8010-0169 HEWL8010-0171 HEWL8010-0173 HEWL8010-0174
Autosampler syringes for CTC PAL3 systems
Spuiten Injectiespuiten voor chromatografie
These syringes are designed to withstand temperatures upto 150 °C.
Syringe specifications are matched to CTC holders to ensure proper fit and reliable functioning.
Used for CTC PAL3 systems (LSI, RSI 120/RSI 85 and RTC 120).