1-Chloro-2-fluoro-5-iodobenzene

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BTB03598.1 BTB03598.10
MAYBBTB03598.1EA 0 EUR
MAYBBTB03598.1 MAYBBTB03598.10
1-Chloro-2-fluoro-5-iodobenzene
1-Chloro-2-fluoro-5-iodobenzene
Formule: C₆H₃ClFI
MDL Nummer: MFCD00042210
CAS nummer: 156150-67-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR