5-Chloro-2-fluorophenyl isothiocyanate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SB01971DA SB01971EA
MAYBSB01971DAEA 0 EUR
MAYBSB01971DA MAYBSB01971EA
5-Chloro-2-fluorophenyl isothiocyanate
5-Chloro-2-fluorophenyl isothiocyanate
Formule: C₇H₃ClFNS
CAS nummer: 247170-25-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR