2-Chloorfenylisocyanaat ≥98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B21489.06 B21489.14
B21489.06EA 44.8 EUR
B21489.06 B21489.14
2-Chloorfenylisocyanaat ≥98%
2-Chloorfenylisocyanaat
Formule: C₇H₄ClNO
MW: 153.57 g/mol
Kookpunt: 189
Dichtheid: 1.273 g/cm³ (20 °C)
CAS nummer: 3320-83-0
EINECS: 222-023-7
UN: 3382
ADR: 6.1 I