2-Chloorfenylisocyanaat
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SB01818.1 SB01818.10 SB01818.50
MAYBSB01818.1EA 0 EUR
MAYBSB01818.1 MAYBSB01818.10 MAYBSB01818.50
2-Chloorfenylisocyanaat
2-Chloorfenylisocyanaat
Formule: C₇H₄ClNO
MW: 153.57 g/mol
Kookpunt: 189
Dichtheid: 1.273 g/cm³ (20 °C)
CAS nummer: 3320-83-0
EINECS: 222-023-7
UN: 3382
ADR: 6.1 I