3,5-Dichlorophenyl isothiocyanate
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
SB01603.1 SB01603.50
MAYBSB01603.1EA 0 EUR
MAYBSB01603.1 MAYBSB01603.50
3,5-Dichlorophenyl isothiocyanate
3,5-Dichlorophenyl isothiocyanate
Formule: C₇H₃Cl₂NS
Kookpunt: 90…100 °C (1 mmHg)
MDL Nummer: MFCD00041072
CAS nummer: 6590-93-8
UN: 2923
ADR: 8,III