3-Chlorophenyl isothiocyanate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

TL00490DA TL00490.10 TL00490.50
MAYBTL00490DAEA 0 EUR
MAYBTL00490DA MAYBTL00490.10 MAYBTL00490.50
3-Chlorophenyl isothiocyanate
3-Chlorophenyl isothiocyanate
Formule: C₇H₄ClNS
Kookpunt: 64…65 °C (0,4 mmHg)
Smeltpunt: 45 °C
Dichtheid: 1,281 g/cm³ (20 °C)
MDL Nummer: MFCD00004805
CAS nummer: 2392-68-9
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR