4-Chlorophenyl isothiocyanate
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
4-Chlorophenylisothiocyanate, 1-Chloro-4-isothiocyanatobenzene
TL00545.1
TL00545.10
TL00545.50
MAYBTL00545.1EA
0
EUR
MAYBTL00545.1
MAYBTL00545.10
MAYBTL00545.50
4-Chlorophenyl isothiocyanate
4-Chlorophenyl isothiocyanate
Formule:
C₇H₄ClNS Kookpunt: 135…142 °C (22 mmHg) Smeltpunt: 46 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00004810 CAS nummer: 2131-55-7 EINECS: 218-358-3 UN: 2923 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...