2,4-Dichlorobenzenesulphonyl chloride
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 2,4-Dichlorobenzene-1-sulphonyl chloride
TL00290.1 TL00290.10 TL00290.50
MAYBTL00290.1EA 0 EUR
MAYBTL00290.1 MAYBTL00290.10 MAYBTL00290.50
2,4-Dichlorobenzenesulphonyl chloride
2,4-Dichlorobenzenesulphonyl chloride
Formule: C₆H₃Cl₃O₂S
Smeltpunt: 56 °C
MDL Nummer: MFCD00052712
CAS nummer: 16271-33-3
UN: 3261
ADR: 8,II