3,5-Dichlorobenzenesulphonyl chloride
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BTB05586.10 BTB05586.50
MAYBBTB05586.10EA 0 EUR
MAYBBTB05586.10 MAYBBTB05586.50
3,5-Dichlorobenzenesulphonyl chloride
3,5-Dichlorobenzenesulphonyl chloride
Formule: C₆H₃Cl₃O₂S
Kookpunt: 83…84 °C (0,1 mmHg)
Smeltpunt: 30…32 °C
MDL Nummer: MFCD00051698
CAS nummer: 705-21-5
UN: 3261
ADR: 8,II