2-Anisoyl chloride
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
SPB06279.1 SPB06279.10 SPB06279.50
MAYBSPB06279.1EA 0 EUR
MAYBSPB06279.1 MAYBSPB06279.10 MAYBSPB06279.50
2-Anisoyl chloride
2-Anisoyl chloride
Formule: C₈H₇ClO₂
Kookpunt: 137 °C (1013 hPa)
Dichtheid: 1,57 g/cm³ (20 °C)
MDL Nummer: MFCD00000664
CAS nummer: 21615-34-9
EINECS: 244-477-5
UN: 1729
ADR: 8,II