3-Anisoyl chloride 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
212970050 212970250 212971000
ACRO212970050EA 31.4 EUR
ACRO212970050 ACRO212970250 ACRO212971000
3-Anisoyl chloride 99%
3-Anisoyl chloride
Formule: C₈H₇ClO₂
Kookpunt: 123…125 °C (15 mmHg)
Dichtheid: 1,214 g/cm³
Vlampunt: 92 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000673
CAS nummer: 1711-05-3
EINECS: 216-975-2
UN: 1729
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5575 to 1.5595 (20°C, 589 nm)
GC ≥98.5 %
Titration RCOCl ≥98.5 %