3,4-Dimethoxybenzoyl chloride
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
KM07765.1 KM07765.10 KM07765.50
MAYBKM07765.1EA 0 EUR
MAYBKM07765.1 MAYBKM07765.10 MAYBKM07765.50
3,4-Dimethoxybenzoyl chloride
3,4-Dimethoxybenzoyl chloride
Formule: C₉H₉ClO₃
Kookpunt: 95…98 °C (1 mmHg)
Smeltpunt: 71 °C
MDL Nummer: MFCD00000674
CAS nummer: 3535-37-3
EINECS: 222-568-0
UN: 3261
ADR: 8,II