Benzanilide 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: N-Phenylbenzamide
148550050 148552500 148550010 148550500
ACRO148550050EA 21.7 EUR
ACRO148550050 ACRO148552500 ACRO148550010 ACRO148550500
Benzanilide 98%
Benzanilide
Formule: C₁₃H₁₁NO
Kookpunt: 117 °C (10 mmHg)
Smeltpunt: 161…165 °C
Dichtheid: 1,32 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003069
CAS nummer: 93-98-1
EINECS: 202-292-7
Merck Index: 13,01060
Order Now
Specification Test Results
Appearance Yellow-green to grey Powder
Infrared spectrum Conforms
Melting point 161 to 165 °C
HPLC ≥97.5 %