2,3-Dimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
D0630-25G
D0630-500G
TCIAD0630-25GEA
58.3
EUR
TCIAD0630-25G
TCIAD0630-500G
2,3-Dimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
2,3-Dimethoxybenzoic acid
Formule:
C₉H₁₀O₄ Kookpunt: 314 °C (1013 hPa) Smeltpunt: 121…124 °C Dichtheid: 1,387 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00002432 CAS nummer: 1521-38-6 EINECS: 216-188-4 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 122.0 to 125.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...