2-(Methylthio)benzoic acid

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

KM06037.1 KM06037.10 KM06037.50
MAYBKM06037.1EA 0 EUR
MAYBKM06037.1 MAYBKM06037.10 MAYBKM06037.50
2-(Methylthio)benzoic acid
2-(Methylthio)benzoic acid
Formule: C₈H₈O₂S
MDL Nummer: MFCD00029934
CAS nummer: 3724-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR