4-Fluorophenylboronic acid 97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: (4-Fluorophenyl)metaboric acid, 4-Fluorobenzeneboronic acid, Dihydroxy(4-fluorophenyl)borane
334100010 334100050 AC33410.1 AC33410.10 334100250
ACRO334100010EA 16.4 EUR
ACRO334100010 ACRO334100050 MAYBAC33410.1 MAYBAC33410.10 ACRO334100250
4-Fluorophenylboronic acid 97%
4-Fluorophenylboronic acid
Formule: C₆H₆BFO₂
MW: 139.92 g/mol
Smeltpunt: 255…259 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00039136
CAS nummer: 1765-93-1
Order Now
Specification Test Results
Appearance White to yellow to beige Powder
Infrared spectrum Conforms
HPLC ≥96.0 %