Membrane filters, mixed cellulose ester, black discs
Leverancier: Advantech
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A045N047A A045R047W
ADVTA045N047AEA 338 EUR
ADVTA045N047A ADVTA045R047W
Membrane filters, mixed cellulose ester, black discs
Filters Membraanfilters
Standard membranes for many laboratory applications including filter sterilising biological fluids, microbiology, contamination analysis and air monitoring.
  • High porosity provides superior flow rate
  • High purity and triton-free
  • Rapid wetting time
  • Non reactive to pyrogens
These membranes are composed of cellulose acetate and cellulose nitrate. These filters withstand autoclaving temperatures up to 130 °C without adversely affecting bubble point, flow rate or microbiological recovery.
Order Now
Specificaties
Environmentally Preferable
Poriëngrootte 0,45 µm
Dikte 135 µm
Waterdebiet 50 ml/min/cm²
Luchtdebiet 5,0 l/min/cm²
Maximumtemperatuur 130 °C
Porositeit 78%