2,5-Difluorophenyl isocyanate ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
L15996.03
L15996.MD
L15996.03EA
39.8
EUR
L15996.03
L15996.MD
2,5-Difluorophenyl isocyanate ≥98%
2,5-Difluorophenyl isocyanate
Purity: 98%
CAS No.: 39718-32-6
Molecular Formula: C7H3F2NO
Formula Weight: 155.11
Storage Temperature: 2°C to 8°C
Flash Point: 56°(132°F)
Refractive Index: 1.4940
Boiling Point: 78°/33mm
Beilstein Reference: 2091753
UN No.: 3080
MDL No.: MFCD00013843
Formule:
C₇H₃F₂NO Kookpunt: 78 °C (33 mmHg) Dichtheid: 1,323 Vlampunt: 56 °C (132 °F) Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00013843 CAS nummer: 39718-32-6 UN: 3080 ADR: 6.1,II |