4-(Trifluoromethoxy)phenyl isocyanate ≥97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
1-Isocyanato-4-(trifluoromethoxy)benzene
B20033.03
B20033.06
B20033.14
B20033.03EA
49.1
EUR
B20033.03
B20033.06
B20033.14
4-(Trifluoromethoxy)phenyl isocyanate ≥97%
4-(Trifluoromethoxy)phenyl isocyanate
Purity: 97%
CAS No.: 35037-73-1
Molecular Formula: C8H4F3NO2
Formula Weight: 203.12
Storage Temperature: 2°C to 8°C
Flash Point: 70°(158°F)
Refractive Index: 1.4580
Melting Point: -29°
Boiling Point: 78°/22mm
Beilstein Reference: 977669
UN No.: 2206
MDL No.: MFCD00035702
Formule:
C₈H₄F₃NO₂ MW: 203.12 g/mol Kookpunt: 78 °C (22 mmHg) Smeltpunt: –29 °C Dichtheid: 1.346 Vlampunt: 70 °C (158 °F) |
MDL Nummer:
MFCD00035702 CAS nummer: 35037-73-1 EINECS: 252-328-0 UN: 2206 ADR: 6.1 III |