2-Fluorophenyl isothiocyanate
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SB01716.1 SB01716.10
MAYBSB01716.1EA 0 EUR
MAYBSB01716.1 MAYBSB01716.10
2-Fluorophenyl isothiocyanate
2-Fluorophenyl isothiocyanate
Formule: C₇H₄FNS
MW: 153.18 g/mol
Kookpunt: 54…58 °C (1 mmHg)
Smeltpunt: 24…26 °C
Dichtheid: 1.24 g/cm³ (20 °C)
MDL Nummer: MFCD00004800
CAS nummer: 38985-64-7
EINECS: 254-237-1
UN: 1760
ADR: 8 II