4-(Difluoromethoxy)aniline

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

TL00507.1 TL00507.10
MAYBTL00507.1EA 0 EUR
MAYBTL00507.1 MAYBTL00507.10
4-(Difluoromethoxy)aniline
4-(Difluoromethoxy)aniline
Formule: C₇H₇F₂NO
Kookpunt: 90 °C (1013 hPa)
Dichtheid: 1,286 g/cm³ (20 °C)
MDL Nummer: MFCD00085005
CAS nummer: 22236-10-8
UN: 2810
ADR: 6.1,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR