1-(Difluoromethoxy)-4-nitrobenzene

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

TL00492.1 TL00492.10
MAYBTL00492.1EA 0 EUR
MAYBTL00492.1 MAYBTL00492.10
1-(Difluoromethoxy)-4-nitrobenzene
1-(Difluoromethoxy)-4-nitrobenzene
Formule: C₇H₅F₂NO₃
MDL Nummer: MFCD00085009
CAS nummer: 1544-86-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR