1-Nitro-4-(trifluoromethoxy)benzene

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 4-(Trifluoromethoxy)nitrobenzene

TL00370.1 TL00370.10
MAYBTL00370.1EA 0 EUR
MAYBTL00370.1 MAYBTL00370.10
1-Nitro-4-(trifluoromethoxy)benzene
1-Nitro-4-(trifluoromethoxy)benzene
Formule: C₇H₄F₃NO₃
MW: 207.11 g/mol
MDL Nummer: MFCD00040302
CAS nummer: 713-65-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR