2,3,4-Trifluorobenzenesulphonyl chloride
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TL00605DA TL00605EA TL00605.50
MAYBTL00605DAEA 0 EUR
MAYBTL00605DA MAYBTL00605EA MAYBTL00605.50
2,3,4-Trifluorobenzenesulphonyl chloride
Trifluorobenzenesulphonyl chloride
Formule: C₆H₂ClF₃O₂S
Kookpunt: 59…61 °C (0,1 mmHg)
Dichtheid: 1,64 g/cm³ (20 °C)
MDL Nummer: MFCD00102573
CAS nummer: 175278-08-7
UN: 3265
ADR: 8,II