2,4-Difluorobenzenesulphonyl chloride
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BTB05552.10 BTB05552.50 BTB05552.1
MAYBBTB05552.10EA 0 EUR
MAYBBTB05552.10 MAYBBTB05552.50 MAYBBTB05552.1
2,4-Difluorobenzenesulphonyl chloride
2,4-Difluorobenzenesulphonyl chloride
Formule: C₆H₃ClF₂O₂S
Kookpunt: 84 °C (1013 hPa)
Dichtheid: 1,581 g/cm³ (20 °C)
MDL Nummer: MFCD00042481
CAS nummer: 13918-92-8
UN: 3265
ADR: 8,II