Pentafluorobenzenesulphonyl chloride 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
147110250 AC14711.1 AC14711.10 AC14711.50 147110050
ACRO147110250EA 218 EUR
ACRO147110250 MAYBAC14711.1 MAYBAC14711.10 MAYBAC14711.50 ACRO147110050
Pentafluorobenzenesulphonyl chloride 99%
Pentafluorobenzenesulphonyl chloride
Formule: C₆ClF₅O₂S
Kookpunt: 211 °C (760 mmHg)
Dichtheid: 1,796 g/cm³
Vlampunt: 90 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007427
CAS nummer: 832-53-1
EINECS: 212-620-0
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4780 to 1.4800 (20°C, 589 nm)
GC ≥98.5 %