4-Iodobenzoyl chloride 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: p-Iodobenzoyl chloride
412390250 412390050
ACRO412390250EA 144 EUR
ACRO412390250 ACRO412390050
4-Iodobenzoyl chloride 98%
4-Iodobenzoyl chloride
Formule: C₇H₄ClIO
MW: 266.47 g/mol
Kookpunt: 100 °C (0.7 mmHg)
Smeltpunt: 62…66 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001058
CAS nummer: 1711-02-0
EINECS: 216-974-7
UN: 3261
ADR: 8 III
Order Now
Specification Test Results
Appearance Light yellow to beige to light brown Crystalline powder and/or chunks or solid
Infrared spectrum Conforms
Melting point 62 to 66 °C
GC ≥97.5 %
Acidity ≤0.0002 meq/g
Peroxides ≤0.002 % (as H2O2)