2-Methoxyphenyl isocyanate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SB01871.1 SB01871.50
MAYBSB01871.1EA 0 EUR
MAYBSB01871.1 MAYBSB01871.50
2-Methoxyphenyl isocyanate
2-Methoxyphenyl isocyanate
Formule: C₈H₇NO₂
Kookpunt: 94,7 °C (1013 hPa)
Dichtheid: 1,122 g/cm³ (20 °C)
MDL Nummer: MFCD00002004
CAS nummer: 700-87-8
EINECS: 211-853-5
UN: 2206
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR