Phenyl isothiocyanate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

SB00933.10 SB00933.50
MAYBSB00933.10EA 0 EUR
MAYBSB00933.10 MAYBSB00933.50
Phenyl isothiocyanate
Phenyl isothiocyanate
Formule: C₇H₅NS
Kookpunt: 221 °C (1013 hPa)
Smeltpunt: –21 °C
Dichtheid: 1,13 g/cm³ (20 °C)
Vlampunt: 87 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00004798
CAS nummer: 103-72-0
EINECS: 203-138-1
UN: 2927
ADR: 6.1,II
Merck Index: 12,07450

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR